Assay Details
Gene Symbol: Rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021661.2 | NP_067693.1 | 3 - 4 | 442 | 83 | |
XM_006235801.2 | XP_006235863.1 | 3 - 4 | 3203 | 83 | ||
XM_008762540.1 | XP_008760762.1 | 2 - 3 | 715 | 83 | ||
XM_008762541.1 | XP_008760763.1 | 2 - 3 | 182 | 83 | ||
GenBank mRNA | AF068136.1 | - | 2 - 3 | 150 | 83 | |
BC088141.1 | - | 3 - 4 | 442 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
