Assay Details
Gene Symbol: Mapk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021746.1 | NP_068514.1 | 2 - 3 | 439 | 65 | |
XM_008765781.1 | XP_008764003.1 | 2 - 3 | 168 | 65 | ||
GenBank mRNA | X96488.1 | - | 2 - 3 | 439 | 65 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008765781.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
