Assay Details
Gene Symbol: Birc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021752.2 | NP_068520.2 | 2 - 3 | 1635 | 94 | |
XM_006242494.2 | XP_006242556.1 | 3 - 4 | 2112 | 94 | ||
XM_006242495.2 | XP_006242557.1 | 3 - 4 | 2313 | 94 | ||
GenBank mRNA | AF183431.1 | - | 2 - 3 | 993 | 94 | |
AF190020.1 | - | 2 - 3 | 2007 | 94 | ||
BC062055.1 | - | 2 - 3 | 1635 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
