Assay Details
Gene Symbol: Timp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021989.2 | NP_068824.1 | 2 - 3 | 362 | 90 | |
GenBank mRNA | AJ409332.1 | - | 2 - 3 | 362 | 90 | |
BC084714.1 | - | 2 - 3 | 454 | 90 | ||
L31884.1 | - | 2 - 3 | 445 | 90 | ||
S72594.1 | - | 2 - 3 | 466 | 90 | ||
S82718.1 | - | 2 - 3 | 455 | 90 | ||
U14526.1 | - | 2 - 3 | 445 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
