Assay Details
Gene Symbol: Pcna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022381.3 | NP_071776.1 | 5 - 6 | 771 | 146 | |
GenBank mRNA | BC060570.1 | - | 5 - 6 | 856 | 6891516 | 146 |
FQ219897.1 | - | 5 - 6 | 852 | 146 | ||
FQ220101.1 | - | 5 - 6 | 852 | 146 | ||
FQ220835.1 | - | 5 - 6 | 852 | 146 | ||
FQ225620.1 | - | 4 - 5 | 476 | 146 | ||
FQ226677.1 | - | 5 - 6 | 851 | 146 | ||
FQ227521.1 | - | 5 - 6 | 852 | 146 | ||
FQ229469.1 | - | 5 - 6 | 852 | 146 | ||
FQ229972.1 | - | 5 - 6 | 853 | 146 | ||
FQ230190.1 | - | 5 - 6 | 852 | 146 | ||
FQ233125.1 | - | 5 - 6 | 852 | 146 | ||
FQ233579.1 | - | 5 - 6 | 852 | 146 | ||
FQ233966.1 | - | 5 - 6 | 852 | 146 | ||
FQ234500.1 | - | 5 - 6 | 852 | 146 | ||
FQ234529.1 | - | 5 - 6 | 852 | 146 | ||
FQ234547.1 | - | 5 - 6 | 852 | 146 | ||
FQ234986.1 | - | 5 - 6 | 852 | 146 | ||
FQ235296.1 | - | 5 - 6 | 852 | 146 | ||
Y00047.1 | - | 5 - 6 | 771 | 146 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
