Assay Details
Gene Symbol: Cd151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022523.1 | NP_071968.1 | 183 | 62 | ||
XM_006230576.2 | XP_006230638.1 | 326 | 62 | |||
GenBank mRNA | AF192547.2 | - | 183 | 62 | ||
BC072515.1 | - | 1 - 2 | 152 | 7104419 | 62 | |
FQ222194.1 | - | 203 | 62 |
Gene Symbol: LOC100911730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749017.3 | XP_003749065.1 | 2 - 3 | 243 | 62 | |
XM_006230598.2 | XP_006230660.1 | 2 - 3 | 326 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF192547.2, BC072515.1, NM_022523.1, XM_003749017.3, XM_006230576.2, XM_006230598.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
