Assay Details
Gene Symbol: Zmat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022548.1 | NP_071993.1 | 1 - 2 | 158 | 122 | |
GenBank mRNA | Y13148.1 | - | 1 - 2 | 158 | 122 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_022548.1, Y13148.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
