Assay Details
Gene Symbol: Entpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022587.1 | NP_072109.1 | 7 - 8 | 1146 | 105 | |
GenBank mRNA | U81295.1 | - | 7 - 8 | 1146 | 105 |
Gene Symbol: LOC103689954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760464.1 | XP_008758686.1 | 1082 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
G protein-coupled receptor signaling pathway
metabolic process
purine ribonucleoside diphosphate catabolic process
response to proline
response to gamma radiation
response to auditory stimulus
platelet activation
response to caffeine
response to lipopolysaccharide
response to ATP
negative regulation of dopamine secretion
response to interferon-beta
cellular response to interferon-alpha
response to morphine
protein homooligomerization
cellular response to lipopolysaccharide
cellular response to aluminum ion
cellular response to tumor necrosis factor
response to L-arginine
negative regulation of ATP biosynthetic process
ATP binding
hydrolase activity
ATP hydrolysis activity
nucleoside-diphosphatase activity
nucleoside-triphosphatase activity
identical protein binding
adenosine-diphosphatase activity
G protein-coupled receptor signaling pathway
metabolic process
purine ribonucleoside diphosphate catabolic process
response to proline
response to gamma radiation
response to auditory stimulus
platelet activation
response to caffeine
response to lipopolysaccharide
response to ATP
negative regulation of dopamine secretion
response to interferon-beta
cellular response to interferon-alpha
response to morphine
protein homooligomerization
cellular response to lipopolysaccharide
cellular response to aluminum ion
cellular response to tumor necrosis factor
response to L-arginine
negative regulation of ATP biosynthetic process
ATP binding
hydrolase activity
ATP hydrolysis activity
nucleoside-diphosphatase activity
nucleoside-triphosphatase activity
identical protein binding
adenosine-diphosphatase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P12252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74111 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12252 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12252 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P12252 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1035058 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P214479 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12252 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702145619 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45293 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12518 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38624 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66927 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28178 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97253 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5676 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702145619 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10715261 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715260 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715267 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715263 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715264 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715262 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715265 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10715258 | - | - |
Rat Genome 230 2.0 Array | 1368885_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368885_at | - | - |
Rat Genome 230 2.0 Array | 1368884_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368884_at | - | - |
Affymetrix HT Rat Focus Array | 1368884_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368884_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100029220.rn.1 | TC0100003599.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100029221.rn.1 | TC0100003599.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100029231.rn.1 | TC0100003599.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100029213.rn.1 | TC0100003599.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100029215.rn.1 | TC0100003599.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100029218.rn.1 | TC0100003599.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100029219.rn.1 | TC0100003599.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100029227.rn.1 | TC0100003599.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100029232.rn.1 | TC0100003599.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100029224.rn.1 | TC0100003599.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100029228.rn.1 | TC0100003599.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10715258 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21934602 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374255 | - | - |
RatRef-12 Expression BeadChip | ILMN_69563 | - | - |
