Assay Details
Gene Symbol: Cnbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022598.1 | NP_072120.1 | 2 - 3 | 287 | 60 | |
GenBank mRNA | AB158421.1 | - | 1 - 2 | 122 | 60 | |
AB158422.1 | - | 1 - 2 | 122 | 60 | ||
AF242550.1 | - | 2 - 3 | 173 | 60 | ||
BC062225.1 | - | 2 - 3 | 271 | 6889682 | 60 | |
D45254.1 | - | 2 - 3 | 287 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
