Assay Details
Gene Symbol: Icos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022610.1 | NP_072132.1 | 4 - 5 | 618 | 68 | |
XM_006245038.2 | XP_006245100.1 | 6 - 7 | 867 | 68 | ||
GenBank mRNA | AB023134.1 | - | 4 - 5 | 618 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
