Assay Details
Gene Symbol: Chrna9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022930.1 | NP_075219.1 | 3 - 4 | 458 | 67 | |
GenBank mRNA | AY574257.1 | - | 3 - 4 | 371 | 67 | |
U12336.1 | - | 3 - 4 | 458 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
