Assay Details
Gene Symbol: Aoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236471.2 | XP_006236533.1 | 1 - 2 | 188 | 83 | |
XM_008762920.1 | XP_008761142.1 | 1 - 2 | 190 | 83 | ||
GenBank mRNA | X73911.1 | - | 1 - 2 | 48 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the X73911.1, XM_006236471.2, XM_008762920.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
