Assay Details
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270701.1 | NP_001257630.1 | 4 - 5 | 957 | 122 | |
NM_001270702.1 | NP_001257631.1 | 4 - 5 | 870 | 122 | ||
NM_001270703.1 | NP_001257632.1 | 4 - 5 | 957 | 122 | ||
NM_023021.2 | NP_075410.2 | 4 - 5 | 957 | 122 | ||
XM_006228466.2 | XP_006228528.1 | 4 - 5 | 2717 | 122 | ||
XM_006228467.2 | XP_006228529.1 | 4 - 5 | 2718 | 122 | ||
XM_006228469.1 | XP_006228531.1 | 5 - 6 | 939 | 122 | ||
XM_006228470.2 | XP_006228532.1 | 4 - 5 | 787 | 122 | ||
XM_006228471.1 | XP_006228533.1 | 4 - 5 | 3214 | 122 | ||
XM_008758975.1 | XP_008757197.1 | 5 - 6 | 2612 | 122 | ||
XM_008758976.1 | XP_008757198.1 | 4 - 5 | 1136 | 122 | ||
XM_008758977.1 | XP_008757199.1 | 3 - 4 | 3118 | 122 | ||
XM_008758978.1 | XP_008757200.1 | 4 - 5 | 776 | 122 | ||
GenBank mRNA | AB292802.1 | - | 2 - 3 | 432 | 122 | |
AB292803.1 | - | 4 - 5 | 815 | 122 | ||
AF149250.1 | - | 4 - 5 | 815 | 122 | ||
AF156554.1 | - | 4 - 5 | 957 | 122 | ||
AF190458.1 | - | 4 - 5 | 927 | 122 | ||
AJ133438.2 | - | 4 - 5 | 815 | 122 | ||
BC128736.1 | - | 4 - 5 | 908 | 122 | ||
DQ137429.1 | - | 1 - 2 | 99 | 122 | ||
EU872449.1 | - | 4 - 5 | 930 | 122 | ||
EU872450.1 | - | 4 - 5 | 930 | 122 | ||
EU872451.1 | - | 4 - 5 | 843 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
