Assay Details
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270701.1 | NP_001257630.1 | 5 - 6 | 1071 | 89 | |
NM_001270702.1 | NP_001257631.1 | 5 - 6 | 984 | 89 | ||
NM_001270703.1 | NP_001257632.1 | 5 - 6 | 1071 | 89 | ||
NM_023021.2 | NP_075410.2 | 5 - 6 | 1071 | 89 | ||
XM_006228466.2 | XP_006228528.1 | 5 - 6 | 2831 | 89 | ||
XM_006228467.2 | XP_006228529.1 | 5 - 6 | 2832 | 89 | ||
XM_006228469.1 | XP_006228531.1 | 6 - 7 | 1053 | 89 | ||
XM_006228470.2 | XP_006228532.1 | 5 - 6 | 901 | 89 | ||
XM_006228471.1 | XP_006228533.1 | 5 - 6 | 3328 | 89 | ||
XM_008758975.1 | XP_008757197.1 | 6 - 7 | 2726 | 89 | ||
XM_008758976.1 | XP_008757198.1 | 5 - 6 | 1250 | 89 | ||
XM_008758977.1 | XP_008757199.1 | 4 - 5 | 3232 | 89 | ||
XM_008758978.1 | XP_008757200.1 | 5 - 6 | 890 | 89 | ||
GenBank mRNA | AB292802.1 | - | 3 - 4 | 546 | 89 | |
AB292803.1 | - | 5 - 6 | 929 | 89 | ||
AF149250.1 | - | 5 - 6 | 929 | 89 | ||
AF156554.1 | - | 5 - 6 | 1071 | 89 | ||
AF190458.1 | - | 5 - 6 | 1041 | 89 | ||
AJ133438.2 | - | 5 - 6 | 929 | 89 | ||
BC128736.1 | - | 5 - 6 | 1022 | 8372655 | 89 | |
EU872449.1 | - | 5 - 6 | 1044 | 89 | ||
EU872450.1 | - | 5 - 6 | 1044 | 89 | ||
EU872451.1 | - | 5 - 6 | 957 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
