Assay Details
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270702.1 | NP_001257631.1 | 8 - 9 | 1332 | 68 | |
NM_001270703.1 | NP_001257632.1 | 8 - 9 | 1220 | 68 | ||
NM_023021.2 | NP_075410.2 | 8 - 9 | 1419 | 68 | ||
XM_006228467.2 | XP_006228529.1 | 8 - 9 | 3180 | 68 | ||
XM_006228471.1 | XP_006228533.1 | 7 - 8 | 3477 | 68 | ||
GenBank mRNA | AB292803.1 | - | 8 - 9 | 1078 | 68 | |
AF156554.1 | - | 8 - 9 | 1419 | 68 | ||
BC128736.1 | - | 8 - 9 | 1370 | 8372655 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
