Assay Details
Gene Symbol: Pdgfd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023962.2 | NP_076452.1 | 3 - 4 | 624 | 75 | |
GenBank mRNA | AB052170.2 | - | 3 - 4 | 624 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
platelet-derived growth factor receptor signaling pathway
regulation of peptidyl-tyrosine phosphorylation
positive regulation of cell division
cellular response to amino acid stimulus
platelet-derived growth factor receptor binding
growth factor activity
multicellular organism development
platelet-derived growth factor receptor signaling pathway
regulation of peptidyl-tyrosine phosphorylation
positive regulation of cell division
cellular response to amino acid stimulus
platelet-derived growth factor receptor binding
growth factor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P409493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108574 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P409493 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P409493 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P409493 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P409496 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P409493 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41913 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32940 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79663 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18927 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8502 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65047 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71427 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52696 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16429 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94075 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71215 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10907846 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10907848 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10907847 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10907845 | - | - |
Rat Genome 230 2.0 Array | 1370727_at | - | - |
Rat Genome 230 2.0 Array | 1370727_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370727_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370727_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800000159.rn.1 | TC0800000031.rn.1 | RTA TC |
Rat Expression Set 230 | 1370727_PM_at | - | - |
Rat Expression Set 230 | 1370727_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10907845 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21792408 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352347 | - | - |
RatRef-12 Expression BeadChip | ILMN_59174 | - | - |
