Assay Details
Gene Symbol: Gapdhs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023964.1 | NP_076454.1 | 4 - 5 | 528 | 88 | |
XM_006228790.2 | XP_006228852.1 | 4 - 5 | 898 | 88 | ||
XM_006228791.2 | XP_006228853.1 | 4 - 5 | 759 | 88 | ||
GenBank mRNA | AJ297631.1 | - | 4 - 5 | 528 | 88 | |
BC161847.1 | - | 4 - 5 | 550 | 7112898 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:

Citations (1)
Maternal thyroid hormone: a strong repressor of neuronal nitric oxide synthase in rat embryonic neocortex.
- Authors
- Sinha RA, Pathak A, Mohan V, Bandyopadhyay S, Rastogi L, Godbole MM
- Journal
- Endocrinology - Page No 4396 (Mon Sep 01 00:00:00 EDT 2008)
- Volume (Year)
- 149 (2008)
- Product Usage
- -
