Assay Details
Gene Symbol: Fgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024145.2 | NP_077059.2 | 2 - 3 | 258 | 78 | |
XM_006239079.2 | XP_006239141.1 | 2 - 3 | 190 | 78 | ||
XM_006239080.2 | XP_006239142.1 | 2 - 3 | 190 | 78 | ||
GenBank mRNA | BC062025.1 | - | 2 - 3 | 258 | 5623501 | 78 |
X57018.1 | - | 2 - 3 | 199 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC062025.1, NM_024145.2, X57018.1, XM_006239079.2, XM_006239080.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
