Assay Details
Gene Symbol: Asic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024154.2 | NP_077068.1 | 1 - 2 | 103 | 70 | |
XM_008765795.1 | XP_008764017.1 | 1 - 2 | 41 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_024154.2, XM_008765795.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
