Assay Details
Gene Symbol: Alox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031010.2 | NP_112272.2 | 12 - 13 | 1649 | 61 | |
XM_006246823.2 | XP_006246885.1 | 14 - 15 | 1930 | 61 | ||
XM_006246824.2 | XP_006246886.1 | 13 - 14 | 1825 | 61 | ||
GenBank mRNA | FQ233819.1 | - | 6 - 7 | 778 | 61 | |
L06040.1 | - | 12 - 13 | 1662 | 61 | ||
S69383.1 | - | 12 - 13 | 1649 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
