Assay Details
Gene Symbol: Creb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031017.1 | NP_112279.1 | 3 - 4 | 299 | 87 | |
XM_006245065.2 | XP_006245127.1 | 3 - 4 | 305 | 87 | ||
XM_006245067.2 | XP_006245129.1 | 2 - 3 | 210 | 87 | ||
GenBank mRNA | FQ211925.1 | - | 3 - 4 | 364 | 87 | |
X14788.1 | - | 3 - 4 | 299 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
