Assay Details
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031081.1 | NP_112343.1 | 11 - 12 | 1413 | 78 | |
XM_006246036.2 | XP_006246098.1 | 11 - 12 | 3265 | 78 | ||
GenBank mRNA | Y15748.1 | - | 11 - 12 | 1413 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
type B pancreatic cell development
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
negative regulation of protein kinase activity
hyperosmotic response
epidermal growth factor receptor signaling pathway
positive regulation of phospholipase activity
regulation of endothelial cell migration
negative regulation of cardiac muscle cell apoptotic process
cell migration
peptidyl-threonine phosphorylation
calcium-mediated signaling
negative regulation of transforming growth factor beta receptor signaling pathway
activation of protein kinase B activity
cellular response to insulin stimulus
negative regulation of toll-like receptor signaling pathway
intracellular signal transduction
regulation of I-kappaB kinase/NF-kappaB signaling
regulation of mast cell degranulation
focal adhesion assembly
positive regulation of release of sequestered calcium ion into cytosol
cellular response to epidermal growth factor stimulus
extrinsic apoptotic signaling pathway
positive regulation of protein localization to plasma membrane
protein serine/threonine kinase activity
3-phosphoinositide-dependent protein kinase activity
insulin receptor binding
ATP binding
phospholipase activator activity
kinase activity
protein kinase binding
phospholipase binding
type B pancreatic cell development
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
negative regulation of protein kinase activity
hyperosmotic response
epidermal growth factor receptor signaling pathway
positive regulation of phospholipase activity
regulation of endothelial cell migration
negative regulation of cardiac muscle cell apoptotic process
cell migration
peptidyl-threonine phosphorylation
calcium-mediated signaling
negative regulation of transforming growth factor beta receptor signaling pathway
activation of protein kinase B activity
cellular response to insulin stimulus
negative regulation of toll-like receptor signaling pathway
intracellular signal transduction
regulation of I-kappaB kinase/NF-kappaB signaling
regulation of mast cell degranulation
focal adhesion assembly
positive regulation of release of sequestered calcium ion into cytosol
cellular response to epidermal growth factor stimulus
extrinsic apoptotic signaling pathway
positive regulation of protein localization to plasma membrane
protein serine/threonine kinase activity
3-phosphoinositide-dependent protein kinase activity
insulin receptor binding
ATP binding
phospholipase activator activity
kinase activity
protein kinase binding
phospholipase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P258751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116127 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P258751 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P258751 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P258751 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P258751 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85942 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56285 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86655 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21861 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10740969 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740968 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740973 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740980 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740975 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740981 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740971 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740972 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740976 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740970 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740977 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740979 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740967 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740974 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740978 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10740966 | - | - |
Rat Genome 230 2.0 Array | 1370052_at | - | - |
Rat Genome 230 2.0 Array | 1370052_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370052_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370052_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000020964.rn.1 | TC1000002378.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020965.rn.1 | TC1000002378.rn.1 | RTA TC |
Rat Expression Set 230 | 1370052_at | - | - |
Rat Expression Set 230 | 1370052_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740966 | - | - |
Rat Genome U34 Set | Y15748_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20935256 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359177 | - | - |
RatRef-12 Expression BeadChip | ILMN_55803 | - | - |
