Assay Details
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031139.1 | NP_112401.1 | 9 - 10 | 1469 | 67 | |
GenBank mRNA | AB035647.1 | - | 9 - 10 | 958 | 67 | |
AB035648.1 | - | 9 - 10 | 877 | 67 | ||
AB035649.1 | - | 8 - 9 | 757 | 67 | ||
AB035650.1 | - | 8 - 9 | 911 | 67 | ||
AB035651.1 | - | 6 - 7 | 591 | 67 | ||
AB047556.1 | - | 9 - 10 | 1469 | 67 | ||
BC128735.1 | - | 9 - 10 | 963 | 7190049 | 67 | |
X90823.1 | - | 7 - 8 | 793 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
