Assay Details
Gene Symbol: Psmd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031331.1 | NP_112621.1 | 1 - 2 | 98 | 64 | |
XM_006232924.2 | XP_006232986.1 | 1 - 2 | 103 | 64 | ||
GenBank mRNA | AB017188.1 | - | 1 - 2 | 98 | 64 | |
AF175575.1 | - | 1 - 2 | 39 | 64 | ||
BC060559.1 | - | 1 - 2 | 48 | 64 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB017188.1, AF175575.1, BC060559.1, NM_031331.1, XM_006232924.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
