Assay Details
Gene Symbol: Igf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031511.2 | NP_113699.2 | 1 - 2 | 1052 | 88 | |
GenBank mRNA | X00911.1 | - | 1 - 2 | 1052 | 88 | |
X14834.1 | - | 1 - 2 | 1120 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_031511.2, X00911.1, X14834.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
