Assay Details
Gene Symbol: Nr4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031628.1 | NP_113816.1 | 2 - 3 | 696 | 61 | |
XM_008763703.1 | XP_008761925.1 | 2 - 3 | 2418 | 61 | ||
GenBank mRNA | D38530.1 | - | 2 - 3 | 696 | 61 | |
DQ268830.1 | - | 2 - 3 | 696 | 61 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the D38530.1, DQ268830.1, NM_031628.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
