Assay Details
Gene Symbol: Foxm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031633.3 | NP_113821.2 | 2 - 3 | 590 | 86 | |
XM_006237418.2 | XP_006237480.1 | 2 - 3 | 590 | 86 | ||
XM_006237420.2 | XP_006237482.1 | 2 - 3 | 590 | 86 | ||
XM_006237421.2 | XP_006237483.1 | 2 - 3 | 590 | 86 | ||
GenBank mRNA | CK469502.1 | - | 1 - 2 | 458 | 86 | |
U83112.1 | - | 2 - 3 | 590 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
