Assay Details
Gene Symbol: Map2k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031643.4 | NP_113831.1 | 1 - 2 | 130 | 70 | |
GenBank mRNA | AI603089.1 | - | 1 - 2 | 130 | 70 | |
BC089772.1 | - | 1 - 2 | 124 | 70 | ||
D13341.1 | - | 1 - 2 | 113 | 70 | ||
D14591.1 | - | 1 - 2 | 83 | 70 | ||
X62313.1 | - | 1 - 2 | 120 | 70 | ||
Z16415.1 | - | 1 - 2 | 113 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
