Assay Details
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031665.2 | NP_113853.1 | 6 - 7 | 764 | 142 | |
GenBank mRNA | BC081769.1 | - | 6 - 7 | 764 | 7134641 | 142 |
U56815.1 | - | 6 - 7 | 740 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P10076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10244 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10076 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10076 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P10076 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10076 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P544887 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33453 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19109 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10764806 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764804 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764801 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764803 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764802 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764810 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764805 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764799 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764809 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764807 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764800 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10764798 | - | - |
Rat Genome 230 2.0 Array | 1388556_at | - | - |
Rat Genome 230 2.0 Array | 1369568_at | - | - |
Rat Genome 230 2.0 Array | 1388556_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369568_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388556_at | - | - |
Affymetrix HT Rat Focus Array | 1369568_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369568_at | - | - |
Affymetrix HT Rat Focus Array | 1388556_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300003628.rn.1 | TC1300000524.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003623.rn.1 | TC1300000524.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003625.rn.1 | TC1300000524.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003627.rn.1 | TC1300000524.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003629.rn.1 | TC1300000524.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003631.rn.1 | TC1300000524.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003630.rn.1 | TC1300000524.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003632.rn.1 | TC1300000524.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003624.rn.1 | TC1300000524.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003626.rn.1 | TC1300000524.rn.1 | RTA TC |
Rat Expression Set 230 | 1369568_at | - | - |
Rat Expression Set 230 | 1388556_PM_at | - | - |
Rat Expression Set 230 | 1388556_at | - | - |
Rat Expression Set 230 | 1369568_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764798 | - | - |
Rat Genome U34 Set | U56815_at | - | - |
Rat Genome U34 Set | rc_AA801183_at | - | - |
Rat Genome U34 Set | U56815_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21671756 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369845 | - | - |
RatRef-12 Expression BeadChip | ILMN_56462 | - | - |
