Assay Details
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031775.3 | NP_113963.2 | 2 - 3 | 205 | 68 | |
GenBank mRNA | AF025670.1 | - | 2 - 3 | 223 | 68 | |
BC078785.1 | - | 2 - 3 | 170 | 68 |
Gene Symbol: LOC103689977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233278.2 | XP_006233340.1 | 2 - 3 | 233 | 68 | |
XM_008761476.1 | XP_008759698.1 | 2 - 3 | 232 | 68 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF025670.1, BC078785.1, NM_031775.3, XM_006233278.2, XM_008761476.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
