Assay Details
Gene Symbol: Src | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031977.1 | NP_114183.1 | 3 - 4 | 366 | 75 | |
XM_008762387.1 | XP_008760609.1 | 3 - 4 | 375 | 75 | ||
XM_008762388.1 | XP_008760610.1 | 3 - 4 | 412 | 75 | ||
GenBank mRNA | AF157016.1 | - | 3 - 4 | 366 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
epidermal growth factor receptor signaling pathway
transforming growth factor beta receptor signaling pathway
integrin-mediated signaling pathway
central nervous system development
cell proliferation
response to mechanical stimulus
response to virus
response to acidic pH
positive regulation of gene expression
regulation of epithelial cell migration
positive regulation of platelet-derived growth factor receptor signaling pathway
positive regulation of glucose metabolic process
positive regulation of smooth muscle cell migration
phosphorylation
single organismal cell-cell adhesion
cell migration
peptidyl-serine phosphorylation
peptidyl-tyrosine phosphorylation
regulation of cell-cell adhesion
intracellular estrogen receptor signaling pathway
forebrain development
response to nutrient levels
positive regulation of protein autophosphorylation
activation of protein kinase B activity
negative regulation of protein homooligomerization
cellular response to insulin stimulus
regulation of intracellular estrogen receptor signaling pathway
adherens junction organization
substrate adhesion-dependent cell spreading
intracellular signal transduction
osteoclast development
cellular response to platelet-derived growth factor stimulus
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
response to drug
response to hydrogen peroxide
positive regulation of apoptotic process
negative regulation of apoptotic process
stress fiber assembly
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of protein binding
positive regulation of MAP kinase activity
positive regulation of phosphatidylinositol 3-kinase activity
transcytosis
innate immune response
bone resorption
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of cell adhesion
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of insulin receptor signaling pathway
protein autophosphorylation
neurotrophin TRK receptor signaling pathway
oogenesis
positive regulation of cytokine secretion
progesterone receptor signaling pathway
positive regulation of protein transport
response to mineralocorticoid
response to electrical stimulus
negative regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
negative regulation of mitochondrial depolarization
uterus development
branching involved in mammary gland duct morphogenesis
regulation of cell projection assembly
positive regulation of ERK1 and ERK2 cascade
response to fatty acid
response to interleukin-1
cellular response to lipopolysaccharide
cellular response to peptide hormone stimulus
cellular response to progesterone stimulus
cellular response to fatty acid
cellular response to hypoxia
cellular response to fluid shear stress
cellular response to transforming growth factor beta stimulus
regulation of podosome assembly
positive regulation of podosome assembly
angiotensin-activated signaling pathway involved in heart process
positive regulation of canonical Wnt signaling pathway
positive regulation of intracellular signal transduction
positive regulation of DNA biosynthetic process
regulation of early endosome to late endosome transport
negative regulation of anoikis
negative regulation of extrinsic apoptotic signaling pathway
negative regulation of intrinsic apoptotic signaling pathway
regulation of caveolin-mediated endocytosis
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
protein kinase C binding
receptor binding
insulin receptor binding
protein binding
ATP binding
protein C-terminus binding
kinase activity
enzyme binding
protein kinase binding
protein domain specific binding
heme binding
estrogen receptor binding
protein complex binding
SH2 domain binding
ion channel binding
ephrin receptor binding
cell adhesion molecule binding
phosphoprotein binding
growth factor receptor binding
scaffold protein binding
protein phosphorylation
epidermal growth factor receptor signaling pathway
transforming growth factor beta receptor signaling pathway
integrin-mediated signaling pathway
central nervous system development
cell proliferation
response to mechanical stimulus
response to virus
response to acidic pH
positive regulation of gene expression
regulation of epithelial cell migration
positive regulation of platelet-derived growth factor receptor signaling pathway
positive regulation of glucose metabolic process
positive regulation of smooth muscle cell migration
phosphorylation
single organismal cell-cell adhesion
cell migration
peptidyl-serine phosphorylation
peptidyl-tyrosine phosphorylation
regulation of cell-cell adhesion
intracellular estrogen receptor signaling pathway
forebrain development
response to nutrient levels
positive regulation of protein autophosphorylation
activation of protein kinase B activity
negative regulation of protein homooligomerization
cellular response to insulin stimulus
regulation of intracellular estrogen receptor signaling pathway
adherens junction organization
substrate adhesion-dependent cell spreading
intracellular signal transduction
osteoclast development
cellular response to platelet-derived growth factor stimulus
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
response to drug
response to hydrogen peroxide
positive regulation of apoptotic process
negative regulation of apoptotic process
stress fiber assembly
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of protein binding
positive regulation of MAP kinase activity
positive regulation of phosphatidylinositol 3-kinase activity
transcytosis
innate immune response
bone resorption
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of cell adhesion
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of insulin receptor signaling pathway
protein autophosphorylation
neurotrophin TRK receptor signaling pathway
oogenesis
positive regulation of cytokine secretion
progesterone receptor signaling pathway
positive regulation of protein transport
response to mineralocorticoid
response to electrical stimulus
negative regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
negative regulation of mitochondrial depolarization
uterus development
branching involved in mammary gland duct morphogenesis
regulation of cell projection assembly
positive regulation of ERK1 and ERK2 cascade
response to fatty acid
response to interleukin-1
cellular response to lipopolysaccharide
cellular response to peptide hormone stimulus
cellular response to progesterone stimulus
cellular response to fatty acid
cellular response to hypoxia
cellular response to fluid shear stress
cellular response to transforming growth factor beta stimulus
regulation of podosome assembly
positive regulation of podosome assembly
angiotensin-activated signaling pathway involved in heart process
positive regulation of canonical Wnt signaling pathway
positive regulation of intracellular signal transduction
positive regulation of DNA biosynthetic process
regulation of early endosome to late endosome transport
negative regulation of anoikis
negative regulation of extrinsic apoptotic signaling pathway
negative regulation of intrinsic apoptotic signaling pathway
regulation of caveolin-mediated endocytosis
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
protein kinase C binding
receptor binding
insulin receptor binding
protein binding
ATP binding
protein C-terminus binding
kinase activity
enzyme binding
protein kinase binding
protein domain specific binding
heme binding
estrogen receptor binding
protein complex binding
SH2 domain binding
ion channel binding
ephrin receptor binding
cell adhesion molecule binding
phosphoprotein binding
growth factor receptor binding
scaffold protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P042955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65945 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042955 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P042955 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P042955 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12708 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702154690 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21092 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702154690 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10841634 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841630 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841626 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841629 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841632 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841635 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841625 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841627 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841631 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841628 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841636 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841633 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10841624 | - | - |
Rat Genome 230 2.0 Array | 1387352_at | - | - |
Rat Genome 230 2.0 Array | 1368830_at | - | - |
Rat Genome 230 2.0 Array | 1368830_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387352_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389030_a_at | - | - |
Rat Genome 230 2.0 Array | 1373124_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389030_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1373124_at | - | - |
Affymetrix HT Rat Focus Array | 1389030_a_at | - | - |
Affymetrix HT Rat Focus Array | 1387352_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373124_at | - | - |
Affymetrix HT Rat Focus Array | 1389030_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1387352_at | - | - |
Affymetrix HT Rat Focus Array | 1368830_at | - | - |
Affymetrix HT Rat Focus Array | 1373124_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368830_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300016100.rn.1 | TC0300002174.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300016101.rn.1 | TC0300002174.rn.1 | RTA TC |
Rat Expression Set 230 | 1387352_at | - | - |
Rat Expression Set 230 | 1389030_PM_a_at | - | - |
Rat Expression Set 230 | 1373124_PM_at | - | - |
Rat Expression Set 230 | 1373124_at | - | - |
Rat Expression Set 230 | 1368830_PM_at | - | - |
Rat Expression Set 230 | 1368830_at | - | - |
Rat Expression Set 230 | 1387352_PM_at | - | - |
Rat Expression Set 230 | 1389030_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10841624 | - | - |
Rat Genome U34 Set | rc_AA875131_at | - | - |
Rat Genome U34 Set | rc_AA943307_g_at | - | - |
Rat Genome U34 Set | rc_AA956919_at | - | - |
Rat Genome U34 Set | rc_AI011267_at | - | - |
Rat Genome U34 Set | rc_AA943307_at | - | - |
Rat Genome U34 Set | rc_AI179933_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21614054 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349852 | - | - |
RatRef-12 Expression BeadChip | ILMN_54506 | - | - |
