Assay Details
Gene Symbol: Eif2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032058.2 | NP_114447.2 | 6 - 7 | 871 | 79 | |
GenBank mRNA | BC070506.1 | - | 6 - 7 | 871 | 7097686 | 79 |
FQ232990.1 | - | 6 - 7 | 880 | 79 | ||
U31880.1 | - | 6 - 7 | 874 | 79 | ||
U83914.1 | - | 6 - 7 | 842 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
