Assay Details
Gene Symbol: Gsk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032080.1 | NP_114469.1 | 10 - 11 | 1331 | 71 | |
XM_006248373.2 | XP_006248435.1 | 11 - 12 | 2076 | 71 | ||
GenBank mRNA | FQ231148.1 | - | 5 - 6 | 599 | 71 | |
FQ233111.1 | - | 5 - 6 | 599 | 71 | ||
X53428.1 | - | 10 - 11 | 1306 | 71 | ||
X73653.1 | - | 10 - 11 | 1331 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
