Assay Details
Gene Symbol: Ppm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270619.1 | NP_001257548.1 | 1 - 2 | 391 | 99 | |
NM_001270620.1 | NP_001257549.1 | 2 - 3 | 865 | 99 | ||
NM_033096.2 | NP_149087.1 | 1 - 2 | 391 | 99 | ||
GenBank mRNA | AJ271834.1 | - | 1 - 2 | 384 | 99 | |
AJ271837.1 | - | 1 - 2 | 384 | 99 | ||
BC061986.1 | - | 1 - 2 | 346 | 5599216 | 99 | |
BC081762.1 | - | 2 - 3 | 865 | 7113731 | 99 | |
FN800175.1 | - | 1 - 2 | 391 | 99 | ||
S90449.1 | - | 1 - 2 | 677 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AJ271834.1, AJ271837.1, BC061986.1, BC081762.1, NM_001270619.1, NM_001270620.1, NM_033096.2, S90449.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
