Assay Details
Gene Symbol: Slc25a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053501.2 | NP_445953.1 | 4 - 5 | 512 | 90 | |
XM_006257531.2 | XP_006257593.1 | 3 - 4 | 505 | 90 | ||
XM_006257532.1 | XP_006257594.1 | 4 - 5 | 549 | 90 | ||
XM_006257534.2 | XP_006257596.1 | 4 - 5 | 1445 | 90 | ||
XM_006257535.1 | XP_006257597.1 | 4 - 5 | 367 | 90 | ||
XM_008773530.1 | XP_008771752.1 | 3 - 4 | 510 | 90 | ||
XM_008773531.1 | XP_008771753.1 | 5 - 6 | 402 | 90 | ||
GenBank mRNA | AB028879.1 | - | 3 - 4 | 309 | 90 | |
AF300424.1 | - | 4 - 5 | 513 | 90 | ||
AJ300165.1 | - | 3 - 4 | 318 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
