Assay Details
Gene Symbol: Nudt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053598.1 | NP_446050.1 | 3 - 4 | 366 | 109 | |
GenBank mRNA | AF253473.1 | - | 3 - 4 | 366 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
