Assay Details
Gene Symbol: Smpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053605.1 | NP_446057.1 | 1 - 2 | 160 | 129 | |
XM_008772541.1 | XP_008770763.1 | 1 - 2 | 193 | 129 | ||
GenBank mRNA | AB000215.1 | - | 1 - 2 | 160 | 129 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB000215.1, NM_053605.1, XM_008772541.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
