Assay Details
Gene Symbol: Csnk1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053615.1 | NP_446067.1 | 1 - 2 | 129 | 114 | |
GenBank mRNA | BC127486.1 | - | 355 | 114 | ||
U77582.1 | - | 1 - 2 | 129 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
