Assay Details
Gene Symbol: Glra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053724.3 | NP_446176.3 | 757 | 114 | ||
GenBank mRNA | AJ310838.1 | - | 757 | 114 | ||
M55250.1 | - | 1108 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
