Assay Details
Gene Symbol: Aurkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053749.1 | NP_446201.1 | 2 - 3 | 231 | 108 | |
XM_006246569.2 | XP_006246631.1 | 2 - 3 | 249 | 108 | ||
XM_008767757.1 | XP_008765979.1 | 2 - 3 | 146 | 108 | ||
XM_008767758.1 | XP_008765980.1 | 2 - 3 | 92 | 108 | ||
GenBank mRNA | BC097297.1 | - | 2 - 3 | 149 | 108 | |
D89731.1 | - | 2 - 3 | 231 | 108 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC097297.1, D89731.1, NM_053749.1, XM_006246569.2, XM_008767757.1, XM_008767758.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
