Assay Details
Gene Symbol: Akr1b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053781.2 | NP_446233.2 | 9 - 10 | 929 | 101 | |
XM_006236250.2 | XP_006236312.1 | 10 - 11 | 1279 | 101 | ||
XM_008762755.1 | XP_008760977.1 | 8 - 9 | 1784 | 101 | ||
GenBank mRNA | AF182168.1 | - | 9 - 10 | 1071 | 101 | |
BC086563.1 | - | 9 - 10 | 929 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
