Assay Details
Gene Symbol: Csnk2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053824.2 | NP_446276.1 | 5 - 6 | 501 | 102 | |
XM_006235220.2 | XP_006235282.1 | 4 - 5 | 561 | 102 | ||
XM_006235221.2 | XP_006235283.1 | 4 - 5 | 602 | 102 | ||
GenBank mRNA | BC091130.1 | - | 5 - 6 | 501 | 7367296 | 102 |
L15618.1 | - | 1118 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
