Assay Details
Gene Symbol: Mapk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053842.1 | NP_446294.1 | 8 - 9 | 1259 | 62 | |
GenBank mRNA | M64300.1 | - | 8 - 9 | 1259 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein phosphorylation
apoptotic process
cellular response to DNA damage stimulus
signal transduction
positive regulation of cell population proliferation
response to toxic substance
animal organ morphogenesis
positive regulation of peptidyl-threonine phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
sensory perception of pain
cytosine metabolic process
positive regulation of cell migration
regulation of protein stability
lipopolysaccharide-mediated signaling pathway
response to lipopolysaccharide
regulation of stress-activated MAPK cascade
mammary gland epithelial cell proliferation
intracellular signal transduction
ERBB signaling pathway
response to exogenous dsRNA
response to estrogen
negative regulation of cell differentiation
positive regulation of translation
positive regulation of transcription, DNA-templated
T cell receptor signaling pathway
B cell receptor signaling pathway
regulation of cytoskeleton organization
long-term synaptic potentiation
lung morphogenesis
trachea formation
labyrinthine layer blood vessel development
ERK1 and ERK2 cascade
response to epidermal growth factor
cellular response to organic substance
caveolin-mediated endocytosis
regulation of Golgi inheritance
cellular response to granulocyte macrophage colony-stimulating factor stimulus
regulation of early endosome to late endosome transport
phosphotyrosine residue binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase activity
protein binding
ATP binding
transcription factor binding
RNA polymerase II CTD heptapeptide repeat kinase activity
kinase activity
protein kinase binding
phosphatase binding
mitogen-activated protein kinase kinase kinase binding
MAPK cascade
protein phosphorylation
apoptotic process
cellular response to DNA damage stimulus
signal transduction
positive regulation of cell population proliferation
response to toxic substance
animal organ morphogenesis
positive regulation of peptidyl-threonine phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
sensory perception of pain
cytosine metabolic process
positive regulation of cell migration
regulation of protein stability
lipopolysaccharide-mediated signaling pathway
response to lipopolysaccharide
regulation of stress-activated MAPK cascade
mammary gland epithelial cell proliferation
intracellular signal transduction
ERBB signaling pathway
response to exogenous dsRNA
response to estrogen
negative regulation of cell differentiation
positive regulation of translation
positive regulation of transcription, DNA-templated
T cell receptor signaling pathway
B cell receptor signaling pathway
regulation of cytoskeleton organization
long-term synaptic potentiation
lung morphogenesis
trachea formation
labyrinthine layer blood vessel development
ERK1 and ERK2 cascade
response to epidermal growth factor
cellular response to organic substance
caveolin-mediated endocytosis
regulation of Golgi inheritance
cellular response to granulocyte macrophage colony-stimulating factor stimulus
regulation of early endosome to late endosome transport
phosphotyrosine residue binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase activity
protein binding
ATP binding
transcription factor binding
RNA polymerase II CTD heptapeptide repeat kinase activity
kinase activity
protein kinase binding
phosphatase binding
mitogen-activated protein kinase kinase kinase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P386375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95559 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P386375 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P386375 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P386375 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P386375 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P386374 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29941 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97821 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82021 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42944 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5029 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53506 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56150 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 93204 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10755809 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755807 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755805 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755812 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755806 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755808 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755810 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755811 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755804 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755813 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10755803 | - | - |
Rat Genome 230 2.0 Array | 1369078_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373426_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369078_at | - | - |
Rat Genome 230 2.0 Array | 1373426_at | - | - |
Rat Genome 230 2.0 Array | 1398346_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398346_at | - | - |
Affymetrix HT Rat Focus Array | 1373426_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373426_at | - | - |
Affymetrix HT Rat Focus Array | 1369078_at | - | - |
Affymetrix HT Rat Focus Array | 1369078_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100013380.rn.1 | TC1100002023.rn.1 | RTA TC |
Rat Expression Set 230 | 1373426_PM_at | - | - |
Rat Expression Set 230 | 1369078_at | - | - |
Rat Expression Set 230 | 1398346_PM_at | - | - |
Rat Expression Set 230 | 1398346_at | - | - |
Rat Expression Set 230 | 1373426_at | - | - |
Rat Expression Set 230 | 1369078_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755803 | - | - |
Rat Genome U34 Set | M64300_at | - | - |
Rat Genome U34 Set | rc_AI172050_at | - | - |
Rat Genome U34 Set | rc_AI103916_at | - | - |
Rat Genome U34 Set | M64300_at | - | - |
Rat Genome U34 Set | M64300_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21502436 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349290 | - | - |
RatRef-12 Expression BeadChip | ILMN_63981 | - | - |
