Assay Details
Gene Symbol: Gsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053906.2 | NP_446358.2 | 6 - 7 | 414 | 63 | |
GenBank mRNA | FM035154.1 | - | 6 - 7 | 414 | 63 | |
U73174.1 | - | 541 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
