Assay Details
Gene Symbol: Sept5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053931.3 | NP_446383.3 | 7 - 8 | 1002 | 101 | |
GenBank mRNA | AB027143.1 | - | 7 - 8 | 1002 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
