Assay Details
Gene Symbol: Sds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | J03863.1 | - | 3 - 4 | 378 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
