Assay Details
Gene Symbol: Ppp2r2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053999.2 | NP_446451.1 | 1 - 2 | 310 | 90 | |
GenBank mRNA | BC128731.1 | - | 1 - 2 | 310 | 90 | |
M83297.1 | - | 1 - 2 | 205 | 90 | ||
M83298.1 | - | 1 - 2 | 291 | 90 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC128731.1, M83297.1, M83298.1, NM_053999.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
