Assay Details
Gene Symbol: Sh3bp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054011.1 | NP_446463.2 | 2 - 3 | 211 | 61 | |
GenBank mRNA | AB027562.1 | - | 2 - 3 | 211 | 61 | |
FQ210473.1 | - | 2 - 3 | 338 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P423662 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P398148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16594 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P398148 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P423662 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P398148 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P423662 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P398148 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P423662 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P398148 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P423662 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29987 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104211 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72117 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 54844 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26905 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1952 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10790225 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790220 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790227 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790228 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790218 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790217 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790221 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790226 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790224 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790222 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10838830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790219 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790223 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10790216 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10838828 | - | - |
Rat Genome 230 2.0 Array | 1387294_at | - | - |
Rat Genome 230 2.0 Array | 1387294_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387294_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387294_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600008975.rn.1 | TC1600001178.rn.1 | RTA TC |
Rat Expression Set 230 | 1387294_PM_at | - | - |
Rat Expression Set 230 | 1387294_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10790216 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21367285 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374910 | - | - |
RatRef-12 Expression BeadChip | ILMN_67095 | - | - |
