Assay Details
Gene Symbol: Park7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057143.2 | NP_476484.1 | 1 - 2 | 278 | 72 | |
GenBank mRNA | AF157511.1 | - | 1 - 2 | 278 | 72 | |
AJ007291.1 | - | 2 - 3 | 203 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF157511.1, AJ007291.1, NM_057143.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
