Assay Details
Gene Symbol: Gabrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080586.1 | NP_542153.1 | 8 - 9 | 1224 | 73 | |
GenBank mRNA | X57514.1 | - | 8 - 9 | 1224 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P13076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307629 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155735 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13076 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13076 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P13076 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13076 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20552 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97316 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90756 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90451 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24468 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83634 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10772552 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772548 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772545 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772551 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772547 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772549 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772550 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772553 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772546 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10772544 | - | - |
Rat Genome 230 2.0 Array | 1387706_at | - | - |
Rat Genome 230 2.0 Array | 1387706_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387706_at | - | - |
Affymetrix HT Rat Focus Array | 1387706_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400003746.rn.1 | TC1400000472.rn.1 | RTA TC |
Rat Expression Set 230 | 1387706_at | - | - |
Rat Expression Set 230 | 1387706_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10772544 | - | - |
Rat Genome U34 Set | X57514_at | - | - |
Rat Genome U34 Set | X57514_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21665548 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372358 | - | - |
RatRef-12 Expression BeadChip | ILMN_48992 | - | - |
